4-Bromo-2,3-difluorobenzyl alcohol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Bromo-2,3-difluorophenyl)methanol
PC500691-1G
PC500691-250MG
APOSPC500691-1GEA
348
EUR
APOSPC500691-1G
APOSPC500691-250MG
4-Bromo-2,3-difluorobenzyl alcohol
4-Bromo-2,3-difluorobenzyl alcohol
Formel:
C₇H₅BrF₂O Molecular Weight: 223,015 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD22493515 CAS-Nummer: 162744-55-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...