4-Iodo-3-(trifluoromethyl)benzaldehyde 95%

Lieferant: Apollo Scientific
Achtung

PC500716-1G PC500716-250MG
APOSPC500716-1GEA 763 EUR
APOSPC500716-1G APOSPC500716-250MG
4-Iodo-3-(trifluoromethyl)benzaldehyde 95%
4-Iodo-3-(trifluoromethyl)benzaldehyde
Formel: C₈H₄F₃IO
Molecular Weight: 300,016 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18398126
CAS-Nummer: 1261777-98-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR