Potassium {[2-(trimethylsilyl)ethoxy]methyl}trifluoroborate 95+%

Lieferant: Apollo Scientific
Achtung

PC50074-1G PC50074-5G
APOSPC50074-1GEA 132 EUR
APOSPC50074-1G APOSPC50074-5G
Potassium {[2-(trimethylsilyl)ethoxy]methyl}trifluoroborate 95+%
Potassium {[2-(trimethylsilyl)ethoxy]methyl}trifluoroborate
Formel: C₆H₁₅BF₃OSi·K
Molecular Weight: 238,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11505930
CAS-Nummer: 1027642-28-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR