4-(Trifluoromethoxy)styrene
Lieferant: Apollo Scientific
|
Achtung
|
PC500744-1G
PC500744-250MG
PC500744-5G
APOSPC500744-1GEA
549
EUR
APOSPC500744-1G
APOSPC500744-250MG
APOSPC500744-5G
4-(Trifluoromethoxy)styrene
4-(Trifluoromethoxy)styrene
Formel:
C₉H₇F₃O Molecular Weight: 188,146 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22413864 CAS-Nummer: 1736-08-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...