5-Bromo-2-(trifluoromethyl)benzaldehyde

Lieferant: Apollo Scientific
Achtung

PC500759-1G PC500759-250MG
APOSPC500759-1GEA 304 EUR
APOSPC500759-1G APOSPC500759-250MG
5-Bromo-2-(trifluoromethyl)benzaldehyde
5-Bromo-2-(trifluoromethyl)benzaldehyde
Formel: C₈H₄BrF₃O
Molecular Weight: 253,016 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28140566
CAS-Nummer: 1288998-92-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR