5-Fluorophthalide

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Fluoro-2-benzofuran-1(3H)-one, 5-Fluoroisobenzofuran-1(3H)-one

PC500768-1G PC500768-5G PC500768-250MG
APOSPC500768-1GEA 52.2 EUR
APOSPC500768-1G APOSPC500768-5G APOSPC500768-250MG
5-Fluorophthalide
5-Fluorophthalide
Formel: C₈H₅FO₂
Molecular Weight: 152,123 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09842448
CAS-Nummer: 700-85-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR