2-Bromo-5-fluoroisonicotinaldehyde 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-5-fluoro-4-formylpyridine, 2-Bromo-5-fluoropyridine-4-carboxaldehyde

PC50077-1G PC50077-5G
APOSPC50077-1GEA 37.7 EUR
APOSPC50077-1G APOSPC50077-5G
2-Bromo-5-fluoroisonicotinaldehyde 95+%
2-Bromo-5-fluoroisonicotinaldehyde
Formel: C₆H₃BrFNO
Molecular Weight: 203,997 g/mol
Schmelzpunkt: 40…42 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09800558
CAS-Nummer: 1005291-43-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR