3-Iodo-4-(trifluoromethyl)benzyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-(Bromomethyl)-2-iodo-1-(trifluoromethyl)benzene
PC500779-1G
PC500779-250MG
APOSPC500779-1GEA
963
EUR
APOSPC500779-1G
APOSPC500779-250MG
3-Iodo-4-(trifluoromethyl)benzyl bromide
3-Iodo-4-(trifluoromethyl)benzyl bromide
Formel:
C₈H₅BrF₃I Molecular Weight: 364,929 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18398192 CAS-Nummer: 1261482-51-2 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...