3-(2,2,2-Trifluoroethoxy)aniline

Lieferant: Apollo Scientific
Achtung

PC500812-1G PC500812-5G
APOSPC500812-1GEA 29 EUR
APOSPC500812-1G APOSPC500812-5G
3-(2,2,2-Trifluoroethoxy)aniline
3-(2,2,2-Trifluoroethoxy)aniline
Formel: C₈H₈F₃NO
Molecular Weight: 191,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07432790
CAS-Nummer: 123207-61-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR