2-Chloro-5-(trifluoromethyl)nicotinaldehyde 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-3-formyl-5-(trifluoromethyl)pyridine, 2-Chloro-5-(trifluoromethyl)pyridine-3-carboxaldehyde

PC50082-1G PC50082-250MG PC50082-5G
APOSPC50082-1GEA 149 EUR
APOSPC50082-1G APOSPC50082-250MG APOSPC50082-5G
2-Chloro-5-(trifluoromethyl)nicotinaldehyde 97%
2-Chloro-5-(trifluoromethyl)nicotinaldehyde
Formel: C₇H₃ClF₃NO
Molecular Weight: 209,553 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10696242
CAS-Nummer: 934279-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR