4-(2,2,2-Trifluoroethoxy)bromobenzene

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-4-(2,2,2-trifluoroethoxy)benzene

PC500829-5G PC500829-100G PC500829-25G
APOSPC500829-5GEA 164 EUR
APOSPC500829-5G APOSPC500829-100G APOSPC500829-25G
4-(2,2,2-Trifluoroethoxy)bromobenzene
4-(2,2,2-Trifluoroethoxy)bromobenzene
Formel: C₈H₆BrF₃O
Molecular Weight: 255,032 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07780193
CAS-Nummer: 106854-77-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR