2,6-Difluoro-3-nitropyridine

Lieferant: Apollo Scientific
Achtung

PC50087-1G PC50087-250MG
APOSPC50087-1GEA 350 EUR
APOSPC50087-1G APOSPC50087-250MG
2,6-Difluoro-3-nitropyridine
2,6-Difluoro-3-nitropyridine
Formel: C₅H₂F₂N₂O₂
Molecular Weight: 160,078 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11044323
CAS-Nummer: 58602-02-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR