S-(4-Fluoro-2-nitrophenyl) dimethylcarbamothioate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: S-(4-Fluoro-2-nitrophenyl) dimethylthiocarbamate, 2-[(Dimethylcarbamoyl)sulphanyl]-5-fluoronitrobenzene

PC50102-1G PC50102-250MG
APOSPC50102-1GEA 615 EUR
APOSPC50102-1G APOSPC50102-250MG
S-(4-Fluoro-2-nitrophenyl) dimethylcarbamothioate 97%
S-(4-Fluoro-2-nitrophenyl) dimethylcarbamothioate
Formel: C₉H₉FN₂O₃S
Molecular Weight: 244,243 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25541895
CAS-Nummer: 1820650-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR