2-Nitro-6-(trifluoromethyl)phenylacetic acid

Lieferant: Apollo Scientific
Achtung

PC501105-1G PC501105-250MG
APOSPC501105-1GEA 522 EUR
APOSPC501105-1G APOSPC501105-250MG
2-Nitro-6-(trifluoromethyl)phenylacetic acid
2-Nitro-6-(trifluoromethyl)phenylacetic acid
Formel: C₉H₆F₃NO₄
Molecular Weight: 249,143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13194459
CAS-Nummer: 1214331-77-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR