4-Amino-3-fluoro-5-nitrobenzotrifluoride 95%

Lieferant: Apollo Scientific
Gefahr

PC501142-1G PC501142-250MG PC501142-5G
APOSPC501142-1GEA 74.1 EUR
APOSPC501142-1G APOSPC501142-250MG APOSPC501142-5G
4-Amino-3-fluoro-5-nitrobenzotrifluoride 95%
4-Amino-3-fluoro-5-nitrobenzotrifluoride
Formel: C₇H₄F₄N₂O₂
Molecular Weight: 224.112 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28348476
CAS-Nummer: 1217304-18-1
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR