3-Fluoro-2-nitrobenzyl alcohol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Fluoro-2-nitrophenyl)methanol
PC501154-1G
PC501154-250MG
PC501154-5G
APOSPC501154-1GEA
46.4
EUR
APOSPC501154-1G
APOSPC501154-250MG
APOSPC501154-5G
3-Fluoro-2-nitrobenzyl alcohol 95%
3-Fluoro-2-nitrobenzyl alcohol
Formel:
C₇H₆FNO₃ Molecular Weight: 171,126 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13185620 CAS-Nummer: 1214323-11-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...