2-Fluoro-3-nitrobenzyl bromide

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(bromomethyl)-2-fluoro-3-nitrobenzene

PC501172-1G PC501172-5G PC501172-25G
APOSPC501172-1GEA 77 EUR
APOSPC501172-1G APOSPC501172-5G APOSPC501172-25G
2-Fluoro-3-nitrobenzyl bromide
2-Fluoro-3-nitrobenzyl bromide
Formel: C₇H₅BrFNO₂
Molecular Weight: 234.023 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11110170
CAS-Nummer: 946125-65-1
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR