5-Bromo-3-fluoro-2-nitroanisole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-1-fluoro-3-methoxy-2-nitrobenzene
PC501204-1G
PC501204-25G
APOSPC501204-1GEA
122
EUR
APOSPC501204-1G
APOSPC501204-25G
5-Bromo-3-fluoro-2-nitroanisole
5-Bromo-3-fluoro-2-nitroanisole
Formel:
C₇H₅BrFNO₃ Molecular Weight: 250,022 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20527806 CAS-Nummer: 1137869-91-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...