3-Chloro-4,5-difluorobenzonitrile 97%

Lieferant: Apollo Scientific
Achtung

PC50123-1G PC50123-5G
APOSPC50123-1GEA 154 EUR
APOSPC50123-1G APOSPC50123-5G
3-Chloro-4,5-difluorobenzonitrile 97%
3-Chloro-4,5-difluorobenzonitrile
Formel: C₇H₂ClF₂N
Molecular Weight: 173,547 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18415562
CAS-Nummer: 103879-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR