4'-Bromo-3'-iodo-2,2,2-trifluoroacetophenone 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(4-Bromo-3-iodophenyl)-2,2,2-trifluoroethanone
PC501269-1G
PC501269-250MG
APOSPC501269-1GEA
719
EUR
APOSPC501269-1G
APOSPC501269-250MG
4'-Bromo-3'-iodo-2,2,2-trifluoroacetophenone 95%
4'-Bromo-3'-iodo-2,2,2-trifluoroacetophenone
Formel:
C₈H₃BrF₃IO Molecular Weight: 378,912 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28348550 CAS-Nummer: 1935411-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...