4-Bromo-2-fluoro-6-iodonitrobenzene

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-1-fluoro-3-iodo-2-nitrobenzene

PC501275-1G PC501275-250MG
APOSPC501275-1GEA 480 EUR
APOSPC501275-1G APOSPC501275-250MG
4-Bromo-2-fluoro-6-iodonitrobenzene
4-Bromo-2-fluoro-6-iodonitrobenzene
Formel: C₆H₂BrFINO₂
Molecular Weight: 345,892 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28348556
CAS-Nummer: 1807008-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR