5-Bromo-3-fluoro-2-iodoanisole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-1-fluoro-2-iodo-3-methoxybenzene
PC501280-1G
PC501280-5G
APOSPC501280-1GEA
60.9
EUR
APOSPC501280-1G
APOSPC501280-5G
5-Bromo-3-fluoro-2-iodoanisole
5-Bromo-3-fluoro-2-iodoanisole
Formel:
C₇H₅BrFIO Molecular Weight: 330,921 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22543687 CAS-Nummer: 1394291-47-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...