2-Bromo-5-fluoro-3-nitrobenzyl bromide

Lieferant: Apollo Scientific
Gefahr

PC501328-1G PC501328-250MG
APOSPC501328-1GEA 335 EUR
APOSPC501328-1G APOSPC501328-250MG
2-Bromo-5-fluoro-3-nitrobenzyl bromide
2-Bromo-5-fluoro-3-nitrobenzyl bromide
Formel: C₇H₄Br₂FNO₂
Molecular Weight: 312.919 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28348592
CAS-Nummer: 1807120-19-9
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR