2-Bromo-4-chloro-6-fluorobenzyl alcohol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-Bromo-4-chloro-6-fluorophenyl)methanol
PC50135-1G
PC50135-250MG
APOSPC50135-1GEA
654
EUR
APOSPC50135-1G
APOSPC50135-250MG
2-Bromo-4-chloro-6-fluorobenzyl alcohol 95%
2-Bromo-4-chloro-6-fluorobenzyl alcohol
Formel:
C₇H₅BrClFO Molecular Weight: 239,469 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24848700 CAS-Nummer: 1449008-26-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...