3-Bromo-4-fluorophenyl methyl sulphone

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-1-fluoro-4-(methylsulphonyl)benzene

PC501363-1G PC501363-250MG
APOSPC501363-1GEA 464 EUR
APOSPC501363-1G APOSPC501363-250MG
3-Bromo-4-fluorophenyl methyl sulphone
3-Bromo-4-fluorophenyl methyl sulphone
Formel: C₇H₆BrFO₂S
Molecular Weight: 253,089 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12032287
CAS-Nummer: 959961-65-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR