4-Fluoro-3-(methylsulphonyl)benzyl bromide 95+%
Lieferant: Apollo Scientific
|
Gefahr
|
PC501367-1G
PC501367-250MG
APOSPC501367-1GEA
1440
EUR
APOSPC501367-1G
APOSPC501367-250MG
4-Fluoro-3-(methylsulphonyl)benzyl bromide 95+%
4-Fluoro-3-(methylsulphonyl)benzyl bromide
Formel:
C₈H₈BrFO₂S Molecular Weight: 267,115 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28348618 CAS-Nummer: 1192347-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...