2-Fluoro-4-(methylsulphonyl)benzyl alcohol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-Fluoro-4-methylsulphonylphenyl)methanol
PC501374-1G
PC501374-250MG
PC501374-5G
APOSPC501374-1GEA
69.6
EUR
APOSPC501374-1G
APOSPC501374-250MG
APOSPC501374-5G
2-Fluoro-4-(methylsulphonyl)benzyl alcohol
2-Fluoro-4-(methylsulphonyl)benzyl alcohol
Formel:
C₈H₉FO₃S Molecular Weight: 204,219 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18633090 CAS-Nummer: 1461702-87-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...