4-[(Difluoromethyl)thio]nitrobenzene 95%

Lieferant: Apollo Scientific

PC50139-1G PC50139-250MG
APOSPC50139-1GEA 186 EUR
APOSPC50139-1G APOSPC50139-250MG
4-[(Difluoromethyl)thio]nitrobenzene 95%
4-[(Difluoromethyl)thio]nitrobenzene
Formel: C₇H₅F₂NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00804873
CAS-Nummer: 24933-57-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR