4-Chloro-5-(trifluoromethoxy)benzene-1,2-diamine
Lieferant: Apollo Scientific
|
Achtung
|
PC501404-100MG
PC501404-250MG
APOSPC501404-100MGEA
60.9
EUR
APOSPC501404-100MG
APOSPC501404-250MG
4-Chloro-5-(trifluoromethoxy)benzene-1,2-diamine
4-Chloro-5-(trifluoromethoxy)benzene-1,2-diamine
Formel:
C₇H₆ClF₃N₂O Molecular Weight: 226,584 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23703477 CAS-Nummer: 156425-08-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...