4-(Trifluoroacetyl)benzyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

PC501409-1G PC501409-250MG
APOSPC501409-1GEA 632 EUR
APOSPC501409-1G APOSPC501409-250MG
4-(Trifluoroacetyl)benzyl bromide 95%
4-(Trifluoroacetyl)benzyl bromide
Formel: C₉H₆BrF₃O
Molecular Weight: 267,043 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28348640
CAS-Nummer: 78126-14-4
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR