Methyl 3-fluoro-5-nitrobenzoate

Lieferant: Apollo Scientific
Achtung

PC501414-1G PC501414-25G PC501414-5G
APOSPC501414-1GEA 21.9 EUR
APOSPC501414-1G APOSPC501414-25G APOSPC501414-5G
Methyl 3-fluoro-5-nitrobenzoate
Methyl 3-fluoro-5-nitrobenzoate
Formel: C₈H₆FNO₄
Molecular Weight: 199,136 g/mol
Schmelzpunkt: 50…52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12172991
CAS-Nummer: 883987-72-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR