3-Fluoro-5-nitrobenzyl alcohol 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC501419-1G
PC501419-5G
APOSPC501419-1GEA
140
EUR
APOSPC501419-1G
APOSPC501419-5G
3-Fluoro-5-nitrobenzyl alcohol 95%
3-Fluoro-5-nitrobenzyl alcohol
Formel:
C₇H₆FNO₃ Molecular Weight: 171,126 g/mol Schmelzpunkt: 33 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD13185621 CAS-Nummer: 883987-74-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...