5-Iodo-2-(trifluoromethyl)-1,3-benzoxazole
Lieferant: Apollo Scientific
|
Achtung
|
PC501479-1G
PC501479-250MG
APOSPC501479-1GEA
975
EUR
APOSPC501479-1G
APOSPC501479-250MG
5-Iodo-2-(trifluoromethyl)-1,3-benzoxazole
5-Iodo-2-(trifluoromethyl)-1,3-benzoxazole
Formel:
C₈H₃F₃INO Molecular Weight: 313.015 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28515363 CAS-Nummer: 1929606-72-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...