2,5-Difluoro-1-(trifluoromethoxy)benzene 99%
Lieferant: Apollo Scientific
|
Achtung
|
PC501501-1G
PC501501-250MG
PC501501-5G
APOSPC501501-1GEA
69.6
EUR
APOSPC501501-1G
APOSPC501501-250MG
APOSPC501501-5G
2,5-Difluoro-1-(trifluoromethoxy)benzene 99%
2,5-Difluoro-1-(trifluoromethoxy)benzene
Formel:
C₇H₃F₅O Molecular Weight: 198,09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22580924 CAS-Nummer: 1404195-06-7 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...