Methyl 3-fluoro-4-formylbenzoate
Lieferant: Apollo Scientific
|
Achtung
|
PC501528-10G
PC501528-1G
PC501528-5G
APOSPC501528-10GEA
793
EUR
APOSPC501528-10G
APOSPC501528-1G
APOSPC501528-5G
Methyl 3-fluoro-4-formylbenzoate
Methyl 3-fluoro-4-formylbenzoate
Formel:
C₉H₇FO₃ Molecular Weight: 182.148 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16036860 CAS-Nummer: 74733-25-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...