Methyl 3-amino-2-bromo-5-fluorobenzoate

Lieferant: Apollo Scientific
Achtung

PC501531-1G PC501531-250MG PC501531-5G
APOSPC501531-1GEA 29 EUR
APOSPC501531-1G APOSPC501531-250MG APOSPC501531-5G
Methyl 3-amino-2-bromo-5-fluorobenzoate
Methyl 3-amino-2-bromo-5-fluorobenzoate
Formel: C₈H₇BrFNO₂
Molecular Weight: 248,049 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17276516
CAS-Nummer: 1342063-52-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR