Methyl 3-fluoro-5-(hydroxymethyl)benzoate 95%

Lieferant: Apollo Scientific
Achtung

PC501539-1G PC501539-250MG PC501539-5G
APOSPC501539-1GEA 464 EUR
APOSPC501539-1G APOSPC501539-250MG APOSPC501539-5G
Methyl 3-fluoro-5-(hydroxymethyl)benzoate 95%
Methyl 3-fluoro-5-(hydroxymethyl)benzoate
Formel: C₉H₉FO₃
Molecular Weight: 184,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23709601
CAS-Nummer: 660416-37-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR