2-Bromo-6-fluoro-4-methylaniline
Lieferant: Apollo Scientific
|
Achtung
|
PC501576-1G
PC501576-5G
APOSPC501576-1GEA
183
EUR
APOSPC501576-1G
APOSPC501576-5G
2-Bromo-6-fluoro-4-methylaniline
2-Bromo-6-fluoro-4-methylaniline
Formel:
C₇H₇BrFN Molecular Weight: 204.04 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07780634 CAS-Nummer: 18349-09-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...