2-Fluoro-3-formylbenzoic acid

Lieferant: Apollo Scientific
Achtung

PC501578-1G PC501578-250MG
APOSPC501578-1GEA 275 EUR
APOSPC501578-1G APOSPC501578-250MG
2-Fluoro-3-formylbenzoic acid
2-Fluoro-3-formylbenzoic acid
Formel: C₈H₅FO₃
Molecular Weight: 168.122 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20526383
CAS-Nummer: 1289043-16-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR