4-Fluoro-3-(hydroxymethyl)benzoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC501590-100MG
PC501590-1G
PC501590-250MG
PC501590-5G
APOSPC501590-100MGEA
0
EUR
APOSPC501590-100MG
APOSPC501590-1G
APOSPC501590-250MG
APOSPC501590-5G
4-Fluoro-3-(hydroxymethyl)benzoic acid 95%
4-Fluoro-3-(hydroxymethyl)benzoic acid
Formel:
C₈H₇FO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD17287664 CAS-Nummer: 816449-68-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...