Methyl 4-fluoro-3-formylbenzoate 99%

Lieferant: Apollo Scientific
Achtung

PC501605-1G PC501605-250MG PC501605-5G
APOSPC501605-1GEA 95.7 EUR
APOSPC501605-1G APOSPC501605-250MG APOSPC501605-5G
Methyl 4-fluoro-3-formylbenzoate 99%
Methyl 4-fluoro-3-formylbenzoate
Formel: C₉H₇FO₃
Molecular Weight: 182.148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16036861
CAS-Nummer: 1093865-65-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR