3-Fluoro-5-(hydroxymethyl)benzoic acid 97%

Lieferant: Apollo Scientific
Achtung

PC501619-100MG PC501619-1G PC501619-250MG
APOSPC501619-100MGEA 49.3 EUR
APOSPC501619-100MG APOSPC501619-1G APOSPC501619-250MG
3-Fluoro-5-(hydroxymethyl)benzoic acid 97%
3-Fluoro-5-(hydroxymethyl)benzoic acid
Formel: C₈H₇FO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD20659871
CAS-Nummer: 816449-67-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR