2-Bromo-6-fluoroacridine-9-carbonitrile

Lieferant: Apollo Scientific
Achtung

PC50163-1G PC50163-250MG PC50163-5G
APOSPC50163-1GEA 108 EUR
APOSPC50163-1G APOSPC50163-250MG APOSPC50163-5G
2-Bromo-6-fluoroacridine-9-carbonitrile
2-Bromo-6-fluoroacridine-9-carbonitrile
Formel: C₁₄H₆BrFN₂
Molecular Weight: 301.113 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32066870
CAS-Nummer: 2375008-66-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR