2-Fluoro-6-formylbenzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC501654-1G
PC501654-250MG
PC501654-5G
APOSPC501654-1GEA
594
EUR
APOSPC501654-1G
APOSPC501654-250MG
APOSPC501654-5G
2-Fluoro-6-formylbenzoic acid
2-Fluoro-6-formylbenzoic acid
Exists in equilibrium with cyclised form - 7-fluoro-3-hydroxy-2-benzofuran-1(3H)-one
Formel:
C₈H₅FO₃ Molecular Weight: 168.122 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28357171 CAS-Nummer: 1289266-50-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...