4-Bromo-3-(difluoromethoxy)aniline 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC501694-1G
PC501694-250MG
PC501694-5G
APOSPC501694-1GEA
783
EUR
APOSPC501694-1G
APOSPC501694-250MG
APOSPC501694-5G
4-Bromo-3-(difluoromethoxy)aniline 95%
4-Bromo-3-(difluoromethoxy)aniline
Formel:
C₇H₆BrF₂NO Molecular Weight: 238.029 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18390862 CAS-Nummer: 87967-37-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...