3-Bromo-2,4-difluorobenzyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

PC501703-1G PC501703-250MG
APOSPC501703-1GEA 783 EUR
APOSPC501703-1G APOSPC501703-250MG
3-Bromo-2,4-difluorobenzyl bromide 95%
3-Bromo-2,4-difluorobenzyl bromide
Formel: C₇H₄Br₂F₂
Molecular Weight: 285,911 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28737920
CAS-Nummer: 1805583-89-4
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR