5-Bromo-2-(difluoromethoxy)aniline 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC501704-1G
PC501704-250MG
PC501704-5G
APOSPC501704-1GEA
335
EUR
APOSPC501704-1G
APOSPC501704-250MG
APOSPC501704-5G
5-Bromo-2-(difluoromethoxy)aniline 95%
5-Bromo-2-(difluoromethoxy)aniline
Formel:
C₇H₆BrF₂NO Molecular Weight: 238.029 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15526767 CAS-Nummer: 1258650-63-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...