4-Bromo-2-(difluoromethoxy)aniline
Lieferant: Apollo Scientific
|
Gefahr
|
PC501721-1G
PC501721-5G
PC501721-250MG
APOSPC501721-1GEA
197
EUR
APOSPC501721-1G
APOSPC501721-5G
APOSPC501721-250MG
4-Bromo-2-(difluoromethoxy)aniline
4-Bromo-2-(difluoromethoxy)aniline
Formel:
C₇H₆BrF₂NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD09878213 CAS-Nummer: 1000575-14-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...