2-Bromo-3-chloro-6-fluoroiodobenzene 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-1-chloro-4-fluoro-3-iodobenzene

PC501764-1G PC501764-250MG PC501764-5G
APOSPC501764-1GEA 575 EUR
APOSPC501764-1G APOSPC501764-250MG APOSPC501764-5G
2-Bromo-3-chloro-6-fluoroiodobenzene 95%
2-Bromo-3-chloro-6-fluoroiodobenzene
Formel: C₆H₂BrClFI
Molecular Weight: 335,34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29037266
CAS-Nummer: 1935178-59-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR