4-Bromo-3-(difluoromethoxy)nitrobenzene 95%

Lieferant: Apollo Scientific
Achtung

PC501766-1G PC501766-250MG
APOSPC501766-1GEA 1090 EUR
APOSPC501766-1G APOSPC501766-250MG
4-Bromo-3-(difluoromethoxy)nitrobenzene 95%
4-Bromo-3-(difluoromethoxy)nitrobenzene
Formel: C₇H₄BrF₂NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD15526849
CAS-Nummer: 835633-47-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR